Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Column Protection System | Avantor

Column Protection System

Stainless steel capillaries 0,18 mm I-Ø (for higher flow rates), nuts and metal ferrules
Katalognr. MANA718971
Anbieter:   MACHEREY-NAGEL
Column Protection System
Katalognr. MANA718971
Anbieter:  MACHEREY-NAGEL
Column Protection System
Column Protection System

Stainless steel capillaries 0,18 mm I-Ø (for higher flow rates), nuts and metal ferrules

Preis
75.20 SFr.
Einloggen für Verfügbarkeit
1 * 3 ST
Product Details & Documents

The Column Protection System guard column holder is used for the installation of NUCLEODUR®, NUCLEOSIL®, NUCLEOSHELL® or UNIVERSAL RP guard columns (EC system) at analytical HPLC columns with ¹/₁₆" fittings.

  • Innovative and universal screw-on guard column holder system
  • Ideal protection for your analytical main column
  • Also suitable for ultra-fast HPLC due to minimised void volume
  • Special ferrules - pressure stability up to 1300 bar

It is advisable to replace the guard column if the silver membrane is contaminated (bright or dark discoloration). If the contaminants are colorless, replace the guard column if the pressure rises or the chromatographic performance decreases.

Lieferinformationen: The column protection system includes 1 cartridge holder, 2 replacement capillaries (0,12 mm ID), 3 ferrules (for HPLC columns with particle size >2 μm), 3 ferrules (for HPLC columns with particle size <2 μm), 2 wrenches and 1 user manual.

Technische Daten
  • Bezeichnung:
    Stainless steel capillaries 0,18 mm I-Ø (for higher flow rates), nuts and metal ferrules
Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen..