Connecting kits for CHROMABOND® Flash columns
Product Details & Documents
CHROMABOND® Flash connecting kits allow to use CHROMABOND® Flash RS and BT cartridges as stand-alone system with any pump, detection, fraction collector combination.
Lieferinformationen: CHROMABOND® Flash Starter Kit consists of ¹/₈" PTFE tubing, I.Ø×L (1,5 mm×3 m), 5×¹/₄"-28 PP nuts, 5×¹/₈" ETFE ferrules, 5×¹/₄"-28 nylon unions, 2×¹/₄"-28 PP Luer locks female, 1×¹/₄"-28 PP Luer locks male and 1×¹/₄"-28 PP Luer tip male.
CHROMABOND® Flash Stand Alone Kit consists of 1×¹/₄"-28 PP Luer lock female, 1×¹/₄"-28 PP Luer lock male, 2×¹/₈" ETFE ferrules, 2×¹/₄"-28 nylon unions and 2×¹/₄"-28 PP nuts.
Dokumente
CHROMABOND® Flash Cartridges.pdf Connecting kits for CHROMABOND® Flash columns.pdf
Weitere Produktdetails
Technische Daten
- Bezeichnung:CHROMABOND® Flash Stand Alone Kit
- Seq:18