Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Connecting kits for CHROMABOND® Flash columns | Avantor
Connecting kits for CHROMABOND® Flash columns
CHROMABOND® Flash Stand Alone Kit
Katalognr. MANA732903
Anbieter:   MACHEREY-NAGEL
Connecting kits for CHROMABOND® Flash columns
Katalognr. MANA732903
Anbieter:  MACHEREY-NAGEL
Connecting kits for CHROMABOND® Flash columns
Connecting kits for CHROMABOND® Flash columns
CHROMABOND® Flash Stand Alone Kit
Preis
SFr. 39.40
Einloggen für Verfügbarkeit
1 * 2 SET
Product Details & Documents
CHROMABOND® Flash connecting kits allow to use CHROMABOND® Flash RS and BT cartridges as stand-alone system with any pump, detection, fraction collector combination.
Lieferinformationen: CHROMABOND® Flash Starter Kit consists of ¹/₈" PTFE tubing, I.Ø×L (1,5 mm×3 m), 5×¹/₄"-28 PP nuts, 5×¹/₈" ETFE ferrules, 5×¹/₄"-28 nylon unions, 2×¹/₄"-28 PP Luer locks female, 1×¹/₄"-28 PP Luer locks male and 1×¹/₄"-28 PP Luer tip male.
CHROMABOND® Flash Stand Alone Kit consists of 1×¹/₄"-28 PP Luer lock female, 1×¹/₄"-28 PP Luer lock male, 2×¹/₈" ETFE ferrules, 2×¹/₄"-28 nylon unions and 2×¹/₄"-28 PP nuts.
Technische Daten
  • Bezeichnung:
    CHROMABOND® Flash Stand Alone Kit
  • Seq:
    18
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.