Ersatzteile-Sets
Acquity H SM-FTN performance maintenance kit. Injector valve cartridge (700005236) not included, rotor seal and pod repair kit included, ACQUITY H-Class SM-FTN, 201000234, WATE
Katalognr. CTSA11-3601
Anbieter: SCIENCIX
Direkt zu
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
Technische Daten
- Bezeichnung:Acquity H SM-FTN performance maintenance kit. Injector valve cartridge (700005236) not included, rotor seal and pod repair kit included
- Seq:14
- Erstausrüster:WATE
- Erstausrüster-Äquivalent:201000234
- Modell:ACQUITY H-Class SM-FTN