Labels, hazard warning, electricity
Hazard labels, Polyester, Schwarz auf gelb, 200×173 mm, 1 label/card
Katalognr. BRDY250223
Anbieter: Brady
Direkt zu
Product Details & Documents
These electricity hazard warning labels are made of polyester and are used for prevention of accidents.
- Self-adhesive
- Clear identification of voltage, phases, grounding and insulation
Dokumente
Technische Daten
- Farbe:Schwarz auf gelb
- Bezeichnung:Hazard labels
- Abmessungen:200×173 mm
- Material:Polyester
- Seq:03
- Verpackt:1 label/card