Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Human IL-8 ELISA Kit | Avantor

Human IL-8 ELISA Kit

Human IL-8 ELISA kit, 96 tests
Katalognr. ABCAAB214030-96
Anbieter:   Abcam
Human IL-8 ELISA Kit
Katalognr. ABCAAB214030-96
Anbieter:  Abcam
CAS Nummer:  
Human IL-8 ELISA Kit
Human IL-8 ELISA Kit

Human IL-8 ELISA kit, 96 tests

CAS Nummer:  
Preis
SFr. 769.00
Einloggen für Verfügbarkeit
1 * 96 Tests
Product Details & Documents

Human IL-8 ELISA kit is a single-wash, 90-minute SimpleStep ELISA® used to quantify human interleukin-8 (IL-8) with a sensitivity of 1,8 pg/ml. The assay employs a simple Mix-Wash-Read protocol with just one incubation and wash step. - Colorimetric sandwich ELISA - 450 nm readout; compatible with any standard plate reader - Available in different formats to meet various throughput needs, including 384-well plates for higher throughput - Broad sample compatibility – suitable for serum, plasma, cell culture supernatants, and cerebrospinal fluid - Cited in over 30 publications.

Technische Daten

Technische Daten

  • Bezeichnung:
    Human IL-8 ELISA kit
  • Tests pro Kit:
    96 tests
  • Empfindlichkeit:
    1,8 pg/ml
  • Reaktivität der primären Antikörper:
    Human
  • Testdauer:
    One step
  • Time to Results:
    1 h 30 min
  • Cat. No.:
    ABCAAB214030-96
  • Lagertemperatur:
    4 °C
  • Format:
    Pre-coated
  • Target protein:
    IL-8
  • Erkennungsmethode:
    Colorimetric
  • Testtyp:
    Sandwich
  • Probentyp:
    Cerebral Spinal Fluid, Heparin Plasma, Citrate plasma, Cell culture supernatant, Serum, EDTA Plasma
  • Erfassungsbereich:
    3,91 - 250 pg/ml
  • Regulatory status:
    RUO

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.