Sucralose NF
Glass, amber, 500 g
Katalognr. SPCMS1416-500GM
Anbieter: Spectrum Chemical
Direkt zu
Product Details & Documents
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Synonyme: Trichlorosucrose, (2R,3R,4R,5R,6R)-2-(((2R,3S,4S,5S)-2,5-Bis(chloromethyl)-3,4-dihydroxytetrahydrofuran-2-yl)oxy)-5-chloro-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4-diol, 1-(1,6-Dichloro-1,6-dideoxy-beta-D-fructofuranosyl)-4-chloro-4-deoxy-alpha-D-galactopyranoside
- CAS Nummer:56038-13-2
- Formel:C₁₂H₁₉Cl₃O₈
- MDL-Nummer:MFCD03648615
- Molekulargewicht:397.63 g/mol
- Schmelzender Platingehalt:130 °C
- Lagertemperatur:Raumtemperatur
Documents
SPCMS1416-2.5KGBL_TDS_EN
Product specification
Technische Daten
Ergebnisse des Spezifikationstests
- APPEARANCE:CONFORM
- ASSAY (ANHYDROUS BASIS):98.0 - 102.0 %
- SPECIFIC ROTATION:+84.0° to +87.5°
- WATER DETERMINATION:Max. 2.0 %
- RESIDUE ON IGNITION:Max. 0.7 %
- CADMIUM (Cd):AS REPORTED
- LEAD (Pb):AS REPORTED
- ARSENIC (As):AS REPORTED
- MERCURY (Hg):AS REPORTED
- HYDROLYSIS PRODUCTS:Max. 0.1 %
- METHANOL:Max. 0.1 %
- RELATED COMPOUNDS:Max. 0.5 %
- IDENTIFICATION A (FTIR):SPECTRUM MATCHES REFERENCE
- IDENTIFICATION (B):RETENTION TIME CORRESPONDS TO STANDARD
- IDENTIFICATION (C):Rf VALUE CORRESPONDS TO STANDARD
- RESIDUAL SOLVENTS:AS REPORTED
Technische Daten
- Packung:Glass, amber
- VE:500 g



