5-Chloranthranilsäure 98%
Bottle, 25 g
Katalognr. APOSOR26820-25G
Anbieter: Apollo Scientific
5-Chloranthranilsäure 98%
Katalognr. APOSOR26820-25G
Anbieter: Apollo Scientific
CAS Nummer: 635-21-2
Direkt zu
Product Details & Documents
Synonyme: 2-Amino-5-chlorobenzoic acid
- CAS Nummer:635-21-2
- Formel:C₇H₆ClNO₂
- MDL-Nummer:MFCD00007838
- Molekulargewicht:171.58 g/mol
- Schmelzender Platingehalt:208…213 °C (Zers.)
- Schmelzender Platingehalt:208…213 °C (Zersetzung)
- Lagertemperatur:Raumtemperatur
- EINECS:211-230-8
Technische Daten
Technische Daten
- Packung:Bottle
- VE:25 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



