Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
4-Ethinylanisol 98%
4-Ethinylanisol 98%
Bottle, 25 g
  APOSOR21951-25G
4-Ethinylanisol 98%
  APOSOR21951-25G
 :  768-60-5
4-Ethinylanisol 98%
Bottle, 25 g
 :  768-60-5
SFr. 67.60
1 * 25 g
 :  (4-Methoxyphenyl)ethyne, (4-Methoxyphenyl)acetylene, 1-Ethynyl-4-methoxybenzene, 1-Methoxy-4-ethynylbenzene
  •  :768-60-5
  •  :C₉H₈O
  •  :MFCD00168815
  •  :132.16 g/mol
  •  :87…91 °C/11mmHg
  •  :28…29 °C
  •  :1,019 g/cm³ (25 °C)
  •  :82 °C
  •  :Raumtemperatur
Documents
  • Packung:
    Bottle
  • VE:
    25 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.