4-Methoxyanthranilsäure
Bottle, 5 g
Catalog # APOSOR17403-5G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: 2-Amino-4-methoxybenzoic acid
- CAS Number:4294-95-5
- Formula:C₈H₉NO₃
- MDL Number:MFCD00667729
- Molecular Weight:167.16 g/mol
- Melting Pt:175…180 °C
- Storage Temperature:Raumtemperatur
Specifications
Specifications
- Packung:Bottle
- VE:5 g



