2-Hydroxyisonicotinic acid
Bottle, 5 g
Katalognr. APOSOR1910-5G
Anbieter: Apollo Scientific
2-Hydroxyisonicotinic acid
Katalognr. APOSOR1910-5G
Anbieter: Apollo Scientific
CAS Nummer: 22282-72-0
Direkt zu
Product Details & Documents
Synonyme: 2-Hydroxy-4-pyridinecarboxylic acid, 2-Hydroxy-isonicotinic acid
- CAS Nummer:22282-72-0
- Formel:C₆H₅NO₃
- MDL-Nummer:MFCD01318566
- Molekulargewicht:139.11 g/mol
- Schmelzender Platingehalt:>300 °C
- Schmelzender Platingehalt:>300 °C
- Lagertemperatur:Raumtemperatur
Technische Daten
Technische Daten
- Packung:Bottle
- VE:5 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



