Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Phthalaldehyd 98%

Phthalaldehyd 98%

Bottle, 25 g
Catalog # APOSOR18349-25G
Corrosive
Toxic
Environment
Supplier:   Apollo Scientific
Phthalaldehyd 98%
Catalog # APOSOR18349-25G
Supplier:  Apollo Scientific
CAS Number:  643-79-8
Phthalaldehyd 98%

Bottle, 25 g

CAS Number:  643-79-8
Price
SFr. 34.70
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde
  • CAS Number:643-79-8
  • Formula:C₈H₆O₂
  • MDL Number:MFCD00003335
  • Molecular Weight:134.13 g/mol
  • Boiling Pt:83 °C/0,8mmHg
  • Melting Pt:53…57 °C
  • Density:1,13 g/cm³ (25 °C)
  • Flash Pt:132 °C
  • Storage Temperature:Kühlschrank
  • EINECS:211-402-2
  • UN:2923
  • ADR:8+6.1,II

Documents

Specifications

Specifications

  • Packung:
    Bottle
  • VE:
    25 g

Specifications