4'-Cyclohexylacetophenon 98%
Bottle, 100 g
Katalognr. APOSOR4954-100G
Anbieter: Apollo Scientific
4'-Cyclohexylacetophenon 98%
Katalognr. APOSOR4954-100G
Anbieter: Apollo Scientific
CAS Nummer: 18594-05-3
Direkt zu
Product Details & Documents
Synonyme: p-Cyclohexylacetophenone
- CAS Nummer:18594-05-3
- Formel:C₁₄H₁₈O
- MDL-Nummer:MFCD00001453
- Molekulargewicht:202.3 g/mol
- Siedepunkt:179…181 °C/14mmHg
- Schmelzender Platingehalt:66…70 °C
- Lagertemperatur:Raumtemperatur
Technische Daten
Technische Daten
- Packung:Bottle
- VE:100 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



