(±)-Isonipecotinsäure
Bottle, 1 kg
Katalognr. APOSOR6923-1KG
Anbieter: Apollo Scientific
Direkt zu
Product Details & Documents
Synonyme: Piperidine-4-carboxylic acid, (±)-4-Piperidinecarboxylic acid, 4-Piperidinecarboxylic acid, Isonipecotic acid
- CAS Nummer:498-94-2
- Formel:C₆H₁₁NO₂
- MDL-Nummer:MFCD00006004
- Molekulargewicht:129.16 g/mol
- Schmelzender Platingehalt:>300 °C
- Schmelzender Platingehalt:>300 °C
- Lagertemperatur:Raumtemperatur
- EINECS:207-872-3
Technische Daten
Technische Daten
- Packung:Bottle
- VE:1 kg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



