Alafosfalin ≥97%
Bottle, 500 mg
Katalognr. APOSBIMI0200-500MG
Anbieter: Apollo Scientific
Direkt zu
Product Details & Documents
Alaphosphin is an Alanine antagonist and can also be used in Murein biosynthesis.
Synonyme: L-Alanyl-L-1-aminoethylphosphonic acid
- CAS Nummer:60668-24-8
- Formel:C₅H₁₃N₂O₄P
- MDL-Nummer:MFCD01569244
- Molekulargewicht:196.141 g/mol
- Lagertemperatur:Kühlschrank
- EINECS:262-362-8
Technische Daten
Technische Daten
- Packung:Bottle
- VE:500 mg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



