L(+)-Cystein
Bottle, 100 g
Catalog # APOSBIC0709-100G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Synonyms: H-Cys-OH, L-(+)-Cysteine, (R)-2-Amino-3-mercaptopropanoic acid
- CAS Number:52-90-4
- Formula:HSCH₂CH(NH₂)CO₂H
- MDL Number:MFCD00064306
- Molecular Weight:121.16 g/mol
- Melting Pt:240 °C (Zersetzung)
- Storage Temperature:Raumtemperatur
Specifications
Specifications
- Packung:Bottle
- VE:100 g



