Agarose
Bottle, 100 g
Katalognr. APOSBIA1176-100G
Anbieter: Apollo Scientific
Direkt zu
Product Details & Documents
- Gel Strength 1.5% >100g/cm2?2200g/cm2
- Gelling temp 36 ± 1.5C
- Melting temp 88 ± 1.5C
- Ash Content <0.45%
- Sulphate <0.1%
- Clarity <4 NTU
- DNA/RNAse Negative.
- Use Rates 0.75% - 2%.
- EEO 0.14 - 0.16
- CAS Nummer:9012-36-6
- MDL-Nummer:MFCD00081294
- Lagertemperatur:Raumtemperatur
- EINECS:232-731-8
Technische Daten
Technische Daten
- Packung:Bottle
- VE:100 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



