Amphotericin B
Bottle, 5 g
Katalognr. APOSBIA0103-5G
Anbieter: Apollo Scientific
Direkt zu
Product Details & Documents
A polyene antifungal antibiotic produced by Streptomyces nodosus.
Synonyme: (1R,3S,5R,6R,9R,11R,15S,16R,17R,18S,19E,21E,23E,25E,27E,29E,31E,33R,35S,36R,37S)-33-(((2R,3S,4S,5S,6R)-4-Amino-3,5-dihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-1,3,5,6,9,11,17,37-octahydroxy-15,16,18-trimethyl-13-oxo-14,39-dioxabicyclo[33.3.1]nonatriaconta-19,21,23,25,27,29,31-heptaene-36-carboxylic acid
- CAS Nummer:1397-89-3
- Formel:C₄₇H₇₃NO₁₇
- MDL-Nummer:MFCD00877763
- Molekulargewicht:924.08 g/mol
- Lagertemperatur:Kühlschrank
Dokumente
Zertifikate
Suche nach ChargennummerTechnische Daten
Technische Daten
- Packung:Bottle
- VE:5 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



