Bicin ≥99%
Bottle, 1 kg
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Biological buffer - useful range 7.6 - 9.0. Recommended for low temperature work.
Synonyms: 2-(Bis(2-hydroxyethyl)amino)acetic acid, N,N-Bis(2-hydroxyethyl)glycine
- CAS Number:150-25-4
- Formula:C₆H₁₃NO₄
- MDL Number:MFCD00004295
- Molecular Weight:163.17 g/mol
- Melting Pt:190 °C
- Storage Temperature:Raumtemperatur
- EINECS:205-755-1
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Packung:Bottle
- VE:1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



