ContactUsLinkComponent
Sulphur ICP Standard Solution 1000 mg/l S 1 * 100 mL
  ROTH2460.1
 :  Roth Carl
Sulphur ICP Standard Solution 1000 mg/l S 1 * 100 mL
  ROTH2460.1
 :  Roth Carl
Frequently Bought Together