Dichtungen
Rotor seal and pod repair kit, vespel, 3-groove for acquity everflow injector pod/cartridge (700002765), ACQUITY UPLC Sample Mgr, ACQUITY Injector Pod (700002765), WATE
Katalognr. CTSA11-4134
Anbieter: SCIENCIX
Direkt zu
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
Technische Daten
- Bezeichnung:Rotor seal and pod repair kit, vespel, 3-groove for acquity everflow injector pod/cartridge (700002765)
- Erstausrüster:WATE
- Erstausrüster-Äquivalent:ACQUITY Injector Pod (700002765)
- Modell:ACQUITY UPLC Sample Mgr