Dichtungen
PEEK™ rotor, HPV, SIL-20 A/C, SIL-20ACHT, SIL-HT, SIL-HTa, SIL-HTc, 228-41310-92, SHIM
CTSACTS-11312
: SCIENCIX
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
- Bezeichnung:PEEK™ rotor, HPV
- Seq:22
- Erstausrüster:SHIM
- Erstausrüster-Äquivalent:228-41310-92
- Modell:SIL-20 A/C, SIL-20ACHT, SIL-HT, SIL-HTa, SIL-HTc