Magnetrührstäbchen
Magnetrührstange, 13 mm, 10 mm
Catalog # VELPA00000357
Supplier: VELP SCIENTIFIC
Jump to:
Product Details & Documents
Certifications:
Accessories information:
Ordering information:
Delivery information:
Specifications
- Ø:10 mm
- Bezeichnung:Magnetrührstange
- Länge:13 mm
- Seq:10
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



