Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Screw caps, 9 mm, Soft-Guard™

Screw caps, 9 mm, Soft-Guard™

9-425 mm, Cross slit (+) Silicone/PTFE, fitted, Gelb
Catalog # MICS9502S-E-3XY
Supplier:   MICROSOLV
Screw caps, 9 mm, Soft-Guard™
Catalog # MICS9502S-E-3XY
Supplier:  MICROSOLV
Screw caps, 9 mm, Soft-Guard™
Screw caps, 9 mm, Soft-Guard™

9-425 mm, Cross slit (+) Silicone/PTFE, fitted, Gelb

Price
SFr. 46.40
Log in for availability
1 * 100 ST
Product Details & Documents

These ribbed screw caps with septa are suitable for 12×32 mm autosampler vials.

Specifications
  • Septum:
    Cross slit (+) Silicone/PTFE, fitted
  • Gewinde:
    9-425 mm
  • Seq:
    07
  • Verschluss-Farbe:
    Gelb
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.