Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Screw caps, 9 mm, Soft-Guard™

Screw caps, 9 mm, Soft-Guard™

9-425 mm, Cross slit (+) Silicone/PTFE, fitted, Natur
Catalog # MICS9502S-E-3XN
Supplier:   MICROSOLV
Screw caps, 9 mm, Soft-Guard™
Catalog # MICS9502S-E-3XN
Supplier:  MICROSOLV
Screw caps, 9 mm, Soft-Guard™
Screw caps, 9 mm, Soft-Guard™

9-425 mm, Cross slit (+) Silicone/PTFE, fitted, Natur

Price
SFr. 46.40
Log in for availability
1 * 100 ST
Product Details & Documents

These ribbed screw caps with septa are suitable for 12×32 mm autosampler vials.

Specifications
  • Septum:
    Cross slit (+) Silicone/PTFE, fitted
  • Gewinde:
    9-425 mm
  • Seq:
    04
  • Verschluss-Farbe:
    Natur