Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Screw caps, 9 mm, Soft-Guard™ | Avantor

Screw caps, 9 mm, Soft-Guard™

9-425 mm, Y- slit Silicone/PTFE, fitted, Blau
Katalognr. MICS9502S-E-34B
Anbieter:   MICROSOLV
Screw caps, 9 mm, Soft-Guard™
Katalognr. MICS9502S-E-34B
Anbieter:  MICROSOLV
Screw caps, 9 mm, Soft-Guard™
Screw caps, 9 mm, Soft-Guard™

9-425 mm, Y- slit Silicone/PTFE, fitted, Blau

Preis
SFr. 46.40
Einloggen für Verfügbarkeit
1 * 100 ST
Product Details & Documents

These ribbed screw caps with septa are suitable for 12×32 mm autosampler vials.

Technische Daten
  • Septum:
    Y- slit Silicone/PTFE, fitted
  • Gewinde:
    9-425 mm
  • Seq:
    22
  • Verschluss-Farbe:
    Blau
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.