Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Instrument trays
300×200×15 mm
Catalog # USBE4762
Supplier:   USBECK Laborgeräte
Instrument trays
Catalog # USBE4762
Instrument trays
Instrument trays
300×200×15 mm
Price
53.20 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Easy to clean
Instrument trays with rounded corner.
Specifications
  • Seq:
    02
  • B×T×H:
    300×200×15 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.