Instrument trays
180×120×10 mm
Katalognr. USBE4760
Anbieter: USBECK Laborgeräte
Direkt zu
Product Details & Documents
Stainless steel.
- Excellent chemical resistance
- Excellent temperature resistance
- Easy to clean
Instrument trays with rounded corner.
Technische Daten
- Seq:01
- B×T×H:180×120×10 mm