Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Ammonia solution 27 - 30% FCC

Ammonia solution 27 - 30% FCC

Glass, amber, 500 mL
Katalognr. SPCMA1293-500MLGL
Ätzend
Reizend
Anbieter:   Spectrum Chemical
Ammonia solution 27 - 30% FCC
Katalognr. SPCMA1293-500MLGL
Anbieter:  Spectrum Chemical
CAS Nummer:  1336-21-6
Ammonia solution 27 - 30% FCC

Glass, amber, 500 mL

CAS Nummer:  1336-21-6
Preis
167.00 SFr.
Einloggen für Verfügbarkeit
1 * 500 mL
Product Details & Documents

Ammonia Solution, FCC is a solution of Ammonia in water. It has many uses, but in food production it is used primarily as a leavening agent. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

GHS hazard : Corrosion;Exclamation mark
UN2672

Synonyme:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Nummer:1336-21-6
  • Formel:NH₃
  • MDL-Nummer:MFCD00066650
  • Molekulargewicht:35.05 g/mol
  • Dichte:0,900
  • Lagertemperatur:Raumtemperatur
  • UN:2672
  • ADR:8,III
Technische Daten

Ergebnisse des Spezifikationstests

  • Assay (NH₃, By Wt.):
    27.0 - 30.0%
  • Lead (Pb):
    max. 0.5 mg/kg
  • Nonvolatile Residue:
    max. 0.02 %
  • Readily Oxidizable Substances:
    To Pass Test
  • Identification:
    To Pass Test

Technische Daten

  • Packung:
    Glass, amber
  • VE:
    500 mL

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.