Instrument trays
215×315×55 mm
Katalognr. USBE4744
Anbieter: USBECK Laborgeräte
Direkt zu
Product Details & Documents
Made of stainless steel.
- Rounded corners
- With rim
- Very good chemical and temperature resistance
Technische Daten
- B×T×H:215×315×55 mm