Dichtungen
Rotor seal, 5-pos. 7-port (1200 bar), 1290, 5068-0005, AGLS
Katalognr. CTSA11-3472
Anbieter: SCIENCIX
Direkt zu
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Sciencix parts are equivalent to the corresponding OEM part.
Technische Daten
- Bezeichnung:Rotor seal, 5-pos. 7-port (1200 bar)
- Seq:53
- Erstausrüster:AGLS
- Erstausrüster-Äquivalent:5068-0005
- Modell:1290