Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Human Recombinant IL-6 | Avantor

Human Recombinant IL-6

100 µG
Katalognr. STMC78050
Marketsource
Human Recombinant IL-6
Katalognr. STMC78050
Marketsource
CAS Nummer:  
Human Recombinant IL-6
Human Recombinant IL-6

100 µG

CAS Nummer:  
Preis
SFr. 519.43
Einloggen für Verfügbarkeit
1 * 100 µG
Product Details & Documents

Interleukin 6 (IL-6) is a pleiotropic growth factor with the wide range of biological activities in immune regulation, hematopoiesis, and oncogenesis. IL-6 is produced by a variety of cell types including T cells, B cells, monocytes and macrophages, fibroblasts, hepatocytes, vascular endothelial cells, and various tumor cell lines. On its own or in combination with other factors such as IL-2 and interferon-γ, IL-6 stimulates the proliferation of B cells, T cells, and hybridoma cells (Hirano et al.; Mihara et al.; Tanaka et al). In combination with cytokines such as IL-3, GM-CSF and SCF, IL-6 has been shown to promote hematopoietic progenitor cell proliferation and differentiation in vitro. IL-6 signals through a cell surface type I cytokine receptor complex consisting of the ligand-binding IL-6α (CD126) and the signal-transducing gp130 subunits. The binding of IL-6 to its receptor system includes activation of JAK/STAT signaling pathway (Mihara et al.; Peters et al.; Tanaka et al.).

Achtung! Bitte beachten: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Technische Daten

Technische Daten

  • VE:
    100 µG
  • Cat. No.:
    STMC78050
  • Purity:
    ≥95%
  • Spezies:
    Human
  • Protein/Peptid-Typ:
    Rekombinant
  • Protein/Peptid-Name:
    IL-6

Technische Daten