Natriumazid 99+%
Plastic bottle, 100 g
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
Synonyme: Sodium triazide
- CAS Nummer:26628-22-8
- Formel:NaN₃
- MDL-Nummer:MFCD00003536
- Molekulargewicht:65.01 g/mol
- Merck-Index:14,08581
- Schmelzender Platingehalt:∼275 °C (Zersetzung)
- Dichte:1,846 g/cm³
- Lagertemperatur:Raumtemperatur
- EINECS:247-852-1
- UN:1687
- ADR:6.1,II
Documents
Technische Daten
Ergebnisse des Spezifikationstests
- Form:White crystalline powder
- Assay (Unspecified):≥99.0%
- Loss on Drying:≤0.5%
Technische Daten
- Packung:Plastic bottle
- VE:100 g
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



