ContactUsLinkComponent
refractometer, rfm742-m, digital, 0-100 brix, 2 d.p, atc, keypad style, factory model 1 * 1 ST
Catalog # BELL19-20-61-182
Supplier:  BELLINGHAM STANLEY
undefined
refractometer, rfm742-m, digital, 0-100 brix, 2 d.p, atc, keypad style, factory model 1 * 1 ST
Catalog # BELL19-20-61-182
Supplier:  BELLINGHAM STANLEY
Frequently Bought Together