Kjeldahl-Tabletten
Product Details & Documents
Kjeldahl-Tabletten sind Katalysatoren, die den Aufschlussschritt der Kjeldahl-Methode zur Bestimmung von Stickstoff fördern.
- Umfangreiches Angebot an Katalysatoren verfügbar
- Verschiedene Tablettengrößen erhältlich
- Das Tablettenformat ist einfach anzuwenden
The Kjeldahl method is used in a wide range of analyses including analysis of proteins, nitrogenated organic materials and also, after reduction, for analysis of nitrates and nitrites. Therefore, the method is used widely in applications from food and beverage testing to agricultural and environmental analysis.
Gesamt-Kjeldahl-Stickstoff (TKN) wird oft als ein Parameter bei Abwasser- und Bodenanalysen herangezogen.
A wide range of catalysts have been developed over the 130 year history of this method to provide ever more rapid and safe digestion methods for particular sets of digestion conditions. Avantor offers a comprehensive range of 'ready to use' tablets to meet the optimal requirements for the sulphuric acid decomposition phase.
Documents
Technische Daten
Technische Daten
- VE:100 Tab.
- Bezeichnung:Kjeldahl Katalysator PS, 5,0 g
- Inhalt:K₂SO₄
Technische Daten
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..