Cyclohexan ≥99,8%, Laborreagens
Glass bottle, 10 L
Anbieter: Honeywell Chemicals
Cyclohexan ≥99,8%, Laborreagens
Katalognr. C100307-4X2.5L
Anbieter: Honeywell Chemicals
CAS Nummer: 110-82-7
Direkt zu
Product Details & Documents
- ≥95% for general laboratory use.
Beilstein Number: 1900225
RTECS Number: GU6300000
UNSPSC Code: 12191504
Synonyme: Hexahydrobenzene, Hexamethylene
- CAS Nummer:110-82-7
- Formel:C₆H₁₂
- MDL-Nummer:MFCD00003814
- Molekulargewicht:84.16 g/mol
- Merck-Index:13,02752
- Siedepunkt:8,3 %(V)
- Schmelzender Platingehalt:6,4 °C
- Dichte:0,78 g/cm³ (20 °C)
- Blitz Pt:–18 °C
- Blitz Pt:-18 °C
- Lagertemperatur:Raumtemperatur
- EINECS:203-806-2
- UN:1145
- ADR:3,II
Dokumente
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance (form):complying
- Appearance (color):complying
- Purity (GC):Min. 99.80 Fl.%
- Non-volatile matter:Max. 0.005 %
- IR spectrum:complying
- Vapor Density:2.9 (vs air)
- Vapor Pressure:77 mmHg ( 20 °C)
- Autoignition Temperature:500 °F
- Refractive Index:n20/D 1.426 (lit.)
Technische Daten
- Packung:Glass bottle
- VE:10 L
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



