Cyclohexan ≥99,8%, Laborreagens
Glass bottle, 1 L
Anbieter: Honeywell Chemicals
Cyclohexan ≥99,8%, Laborreagens
Katalognr. C100307-1L
Anbieter: Honeywell Chemicals
CAS Nummer: 110-82-7
Direkt zu
Product Details & Documents
- ≥95% for general laboratory use.
Beilstein Number: 1900225
RTECS Number: GU6300000
UNSPSC Code: 12191504
Synonyme: Hexahydrobenzene, Hexamethylene
- CAS Nummer:110-82-7
- Formel:C₆H₁₂
- MDL-Nummer:MFCD00003814
- Molekulargewicht:84.16 g/mol
- Merck-Index:13,02752
- Siedepunkt:8,3 %(V)
- Schmelzender Platingehalt:6,4 °C
- Dichte:0,78 g/cm³ (20 °C)
- Blitz Pt:–18 °C
- Blitz Pt:-18 °C
- Lagertemperatur:Raumtemperatur
- EINECS:203-806-2
- UN:1145
- ADR:3,II
Documents
Technische Daten
Ergebnisse des Spezifikationstests
- Appearance (form):complying
- Appearance (color):complying
- Purity (GC):Min. 99.80 Fl.%
- Non-volatile matter:Max. 0.005 %
- IR spectrum:complying
- Vapor Density:2.9 (vs air)
- Vapor Pressure:77 mmHg ( 20 °C)
- Autoignition Temperature:500 °F
- Refractive Index:n20/D 1.426 (lit.)
Technische Daten
- Packung:Glass bottle
- VE:1 L



