Warning signs
Warning of laser beam, Vinyl, selbstklebend, 15×13 mm
Katalognr. BRDY826905
Anbieter: Brady
Direkt zu
Product Details & Documents
These safety signs are for the purposes of accident prevention, fire protection, health hazard information and emergency evacuation.
- Recommended by ISO 7010
- Colour: Yellow/Black
- Shape: Triangle
Technische Daten
- Bezeichnung:Warning of laser beam
- Abmessungen:15×13 mm
- Material:Vinyl, selbstklebend
- Seq:01