Kegelschliffstopfen
24/29, Amber glass
Catalog # BRND144676
Supplier: Brand
Jump to:
Product Details & Documents
DURAN®.Hohl, mit Abtropfspitze und Sechskantgriffscheibe.
Certifications: DIN 12252.
Documents
BRND144635_TDS_EN
Datasheet
Specifications
- NS:24/29
- Typ:Amber glass
- Seq:19
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.