Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
 

Kryoboxen, Karton

8×8, Grau, 14 mm, 130×130×30 mm
  NEOL2-2882
 :   NEOLAB
Kryoboxen, Karton
  NEOL2-2882
 :  NEOLAB
Kryoboxen, Karton

8×8, Grau, 14 mm, 130×130×30 mm

 
28.90 SFr.
   
1 * 1 ST
 

These cryoboxes are made of cryo resistant and impregnated cardboard.

  • Pop-up lid without labelling field

For storage of cryovials.

 
  • Fächer:
    8×8
  • Für Röhrchen Ø:
    14 mm
  • Außen B×T×H:
    130×130×30 mm
  • Farbe:
    Grau
  • Seq:
    08
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.