Kryoboxen, Karton
Weiß, 133×133×50 mm
Katalognr. NEOL2-2671
Anbieter: NEOLAB
Direkt zu
Product Details & Documents
These cryoboxes are made of cryo resistant and impregnated cardboard.
- Pop-up lid without labelling field
For storage of cryovials.
Technische Daten
- Außen B×T×H:133×133×50 mm
- Farbe:Weiß
- Seq:01