Group lockout boxes for wall-mounting
Metal group lockout box, large, for 12 padlocks, Rot, 406×57,15×178 mm
Katalognr. BRDY105715
Anbieter: Brady
Direkt zu
Product Details & Documents
These lockout boxes are made of durable, powder-coated steel with key slot on top.
- Clear Lexan window slides out when locks are removed
- Comes with 7 or 15 key pegs that hold 5 keys each and ID labels to simplify key management
- Pre-drilled holes and hardware included for quick mounting
Legend language: English
Technische Daten
- Typ:Metal group lockout box, large, for 12 padlocks
- Farbe:Rot
- Seq:02
- B×T×H:406×57,15×178 mm