Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
ImmunoCult™-ACF Dendritic Cell Medium | Avantor
ImmunoCult™-ACF Dendritic Cell Medium
ImmunoCult™ ACF dendritic cell medium, 500 mL
Katalognr. STMC10986
Marketsource
ImmunoCult™-ACF Dendritic Cell Medium
Katalognr. STMC10986
Marketsource
CAS Nummer:  
ImmunoCult™-ACF Dendritic Cell Medium
ImmunoCult™-ACF Dendritic Cell Medium
ImmunoCult™ ACF dendritic cell medium, 500 mL
CAS Nummer:  
Preis
SFr. 457.13
Einloggen für Verfügbarkeit
1 * 500 mL
Product Details & Documents
Medium for the culture and differentiation of human monocytes into dendritic cells and their maturation.
Reliably culture and differentiate human monocytes into dendritic cells (DCs) using the serum- and animal component-free (ACF) ImmunoCult™-ACF Dendritic Cell Medium. For your convenience, the medium is available either individually or as part of ImmunoCult™ Dendritic Cell Culture Kit. Use the ImmunoCult™-ACF Dendritic Cell Medium in combination with ImmunoCult™-ACF Dendritic Cell Differentiation Supplement to generate DCs in vitro.
To support the maturation of immature DCs to mature DCs, combine with the ImmunoCult™ Dendritic Cell Maturation Supplement.
Achtung! Bitte beachten: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Technische Daten
Technische Daten
  • VE:
    500 mL
  • Bezeichnung:
    ImmunoCult™ ACF dendritic cell medium
Technische Daten
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..