Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Sunset yellow FCF

Sunset yellow FCF

100 g
Catalog # SPCMFD155-100GM
Supplier:   Spectrum Chemical
Sunset yellow FCF
Catalog # SPCMFD155-100GM
Supplier:  Spectrum Chemical
CAS Number:  2783-94-0
Sunset yellow FCF

100 g

CAS Number:  2783-94-0
Price
94.20 SFr.
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Sodium 6-hydroxy-5-((4-sulphonatophenyl)diazenyl)naphthalene-2-sulphonate, Food yellow No. 5, FD and C Yellow No. 6
  • CAS Number:2783-94-0
  • Formula:C₁₆H₁₀N₂Na₂O₇S₂
  • MDL Number:MFCD00036437
  • Molecular Weight:452.36 g/mol

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • VE:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.