ISIS 3
ISIS valve rotor seal replacement kit, includes rotor seal for ISIS valve and ⁷/₆₄" hex wrench
Katalognr. HEWLG8411-67111
Anbieter: AGILENT HEWLETT PACKARD
Direkt zu
Product Details & Documents
The ISIS 3 dramatically increases the productivity of both the Agilent 7800 ICP-MS and the Agilent 7900 ICP-MS while also allowing high matrix samples to be run for extended periods of time.
- Constant flow nebulisation eliminates stabilisation after high-speed uptake
- Reduces the amount of sample reaching the instrument, reducing cleaning and maintenance requirements
- Provides the highest possible throughput with discrete sampling
With its fast uptake piston pump and close-coupled 7-port valve located close to the nebuliser, the ISIS 3 enables reduced sample uptake and rinse times with discrete sampling (DS). The ISIS 3 delivers unmatched productivity for high sample matrix analysis in high throughput laboratories.
Technische Daten
- Bezeichnung:ISIS valve rotor seal replacement kit, includes rotor seal for ISIS valve and ⁷/₆₄" hex wrench
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..