Graphite platforms for PerkinElmer
Product Details & Documents
These graphite platforms and tubes for PerkinElmer graphite furnace AA systems are quality tested to ensure every batch passes our demanding performance specifications for contamination, sensitivity, precision, electrical resistance, and lifetime.
- Guaranteed to be compatible with PerkinElmer instruments
- Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
- Parts qualified on a genuine PerkinElmer system
L'vov platforms isolate the sample from the tube walls to allow indirect heating of the sample on the platform. The platform heats primarily by the radiation given off from the tube walls. Sample vaporisation and atomisation occurs after the tube reaches a steady-state temperature. L'vov platforms ensure vaporisation occurs into a higher temperature gas atmosphere, reducing interferences. The platforms also provide longer tube life through reduced attack by sample matrix and reagents.
Instruction guides and how-to videos are available to ensure to get the best performance from PerkinElmer system when using parts and supplies.
Technische Daten
- Bezeichnung:L'vov platform, for HGA tubes with internal grooves, for HGA 300/600/700/800/850/900 furnaces
- Seq:02