Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
MP-AES sample introduction kit supplies
MP-AES sample introduction kit supplies
Aqueous sample introduction kit, for Agilent MP-AES
Codice catalogo HEWLG8003-67001
Nome del fornitore:   AGILENT HEWLETT PACKARD
MP-AES sample introduction kit supplies
Codice catalogo HEWLG8003-67001
Nome del fornitore:  AGILENT HEWLETT PACKARD
MP-AES sample introduction kit supplies
MP-AES sample introduction kit supplies
Aqueous sample introduction kit, for Agilent MP-AES
Price
Richiedi prezzo
Log in per disponibilità
1 * 1 ST
Product Details & Documents
MP-AES Sample Kits provide convenient kits for handling aqueous/acidic and organic samples. Select the appropriate one for your application.
  • Agilent nebulisers are manufactured to fine tolerances for reproducible and stable sample uptake
  • Tight specifications deliver fine droplets for best signal intensity and optimum signal to noise
  • Fewer repeats means less downtime and improved productivity
Agilent's spray chambers are designed to provide fine droplets for the best signal intensity and optimum signal to noise performance with excellent washout. The unique OneNeb nebuliser provides improved sensitivity and provides greater tolerance to dissolved solids, strong acids such as HF and resistance to most common organic solvents Featuring Flow Blurring technology, this universal nebuliser provides efficient operation over a much wider flow rate range.
Agilent provides a choice of convenient kits for handling aqueous or acidic samples with the Agilent MP-AES instrument. Each kit includes single pass glass cyclonic spray chamber, nebulizer (concentric for the high sensitivity kit and Conikal for the routine kit) and PVC peristaltic pump tubing with white/white tabs (excludes peristaltic pump tubes for spray chamber drain).
The MP-AES organics kit, includes all items required to adapt the 4100 MP-AES for determination of organic solvents including External Gas Control Module (EGCM), inert OneNeb nebuliser, and solvent resistant tubing (does not include a double pass glass cyclonic spray chamber which is recommended). 4200 MP-AES users should order only the EGCM (p/n G8004A), as all other items are supplied by default.
Specifiche
  • Bezeichnung:
    Aqueous sample introduction kit, for Agilent MP-AES
  • Seq:
    02
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..