Burner system components for PerkinElmer
Burner drain assembly, with tubing and drain interlock, drain bottle not included, for PinAAcle
Catalog # HEWL8003-0857
Supplier: AGILENT HEWLETT PACKARD
Jump to:
Product Details & Documents
Agilent burner components for PerkinElmer AA systems are designed to provide the best precision, efficient drainage, minimal burner blockage, and reduced interferences.
- Guaranteed to be compatible with PerkinElmer instruments
- Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies.
Specifications
- Bezeichnung:Burner drain assembly, with tubing and drain interlock, drain bottle not included, for PinAAcle
- Seq:04