AuftragsverfolgungKontakt
Burner system components for PerkinElmer
Burner system components for PerkinElmer
Burner drain assembly, with tubing and drain interlock, drain bottle not included, for PinAAcle
Catalog # HEWL8003-0857
Burner system components for PerkinElmer
Catalog # HEWL8003-0857
Burner system components for PerkinElmer
Burner drain assembly, with tubing and drain interlock, drain bottle not included, for PinAAcle
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Agilent burner components for PerkinElmer AA systems are designed to provide the best precision, efficient drainage, minimal burner blockage, and reduced interferences.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies.
Specifications
  • Bezeichnung:
    Burner drain assembly, with tubing and drain interlock, drain bottle not included, for PinAAcle
  • Seq:
    04