AuftragsverfolgungKontakt
ISIS 3 | Avantor
ISIS 3
Flangeless tube end fitting, ¹/₁₆", with PP nut with green collapsible PCTFE ferrule to suit ¹/₁₆" tubing
Katalognr. HEWL5023-1525
ISIS 3
Katalognr. HEWL5023-1525
ISIS 3
ISIS 3
Flangeless tube end fitting, ¹/₁₆", with PP nut with green collapsible PCTFE ferrule to suit ¹/₁₆" tubing
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 5 ST
Product Details & Documents
The ISIS 3 dramatically increases the productivity of both the Agilent 7800 ICP-MS and the Agilent 7900 ICP-MS while also allowing high matrix samples to be run for extended periods of time.
  • Constant flow nebulisation eliminates stabilisation after high-speed uptake
  • Reduces the amount of sample reaching the instrument, reducing cleaning and maintenance requirements
  • Provides the highest possible throughput with discrete sampling
With its fast uptake piston pump and close-coupled 7-port valve located close to the nebuliser, the ISIS 3 enables reduced sample uptake and rinse times with discrete sampling (DS). The ISIS 3 delivers unmatched productivity for high sample matrix analysis in high throughput laboratories.
Technische Daten
  • Bezeichnung:
    Flangeless tube end fitting, ¹/₁₆", with PP nut with green collapsible PCTFE ferrule to suit ¹/₁₆" tubing