ISIS 3
Flangeless tube end fitting, ¹/₁₆", with PP nut with black collapsible PCTFE ferrule to suit ¹/₁₆" tubing
Katalognr. HEWL5023-1523
Anbieter: AGILENT HEWLETT PACKARD
Direkt zu
Product Details & Documents
The ISIS 3 dramatically increases the productivity of both the Agilent 7800 ICP-MS and the Agilent 7900 ICP-MS while also allowing high matrix samples to be run for extended periods of time.
- Constant flow nebulisation eliminates stabilisation after high-speed uptake
- Reduces the amount of sample reaching the instrument, reducing cleaning and maintenance requirements
- Provides the highest possible throughput with discrete sampling
With its fast uptake piston pump and close-coupled 7-port valve located close to the nebuliser, the ISIS 3 enables reduced sample uptake and rinse times with discrete sampling (DS). The ISIS 3 delivers unmatched productivity for high sample matrix analysis in high throughput laboratories.
Technische Daten
- Bezeichnung:Flangeless tube end fitting, ¹/₁₆", with PP nut with black collapsible PCTFE ferrule to suit ¹/₁₆" tubing
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..