Quick connect torch modules for PerkinElmer
Torch module, quick change, for HF analysis. Includes GemCone high dissolved solids nebuliser, alumina, 2,0 mm Int.Ø injector, quartz 3-slot torch, HF-resistant baffled cyclonic spray chamber, for Optima 4300 V/5300 V/7300 V
Katalognr. HEWL8003-0342
Anbieter: AGILENT HEWLETT PACKARD
Quick connect torch modules for PerkinElmer
Katalognr. HEWL8003-0342
Anbieter: AGILENT HEWLETT PACKARD
Direkt zu
Product Details & Documents
The Quick-Connect torch module for PerkinElmer ICP-OES includes the torch, mounting block, 2 mm alumina injector, Scott spray chamber, and Cross-Flow nebuliser.
- Guaranteed to be compatible with PerkinElmer instruments
- Backed by the stand behind warranty that guarantees the parts will not cause instrument failure or downtime
- Parts qualified on a genuine PerkinElmer system
- Many parts in packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage
The Quick-Change module is useful when you are running different sample types and need separate sample introduction for each.
Technische Daten
- Bezeichnung:Torch module, quick change, for HF analysis. Includes GemCone high dissolved solids nebuliser, alumina, 2,0 mm Int.Ø injector, quartz 3-slot torch, HF-resistant baffled cyclonic spray chamber, for Optima 4300 V/5300 V/7300 V
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..