AuftragsverfolgungKontakt
Quick connect torch modules for PerkinElmer | Avantor
Quick connect torch modules for PerkinElmer
Torch module, quick change, for HF analysis. Includes GemCone high dissolved solids nebuliser, alumina, 2,0 mm Int.Ø injector, quartz 3-slot torch, HF-resistant baffled cyclonic spray chamber, for Optima 4300 V/5300 V/7300 V
Katalognr. HEWL8003-0342
Quick connect torch modules for PerkinElmer
Katalognr. HEWL8003-0342
Quick connect torch modules for PerkinElmer
Torch module, quick change, for HF analysis. Includes GemCone high dissolved solids nebuliser, alumina, 2,0 mm Int.Ø injector, quartz 3-slot torch, HF-resistant baffled cyclonic spray chamber, for Optima 4300 V/5300 V/7300 V
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
The Quick-Connect torch module for PerkinElmer ICP-OES includes the torch, mounting block, 2 mm alumina injector, Scott spray chamber, and Cross-Flow nebuliser.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the stand behind warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
  • Many parts in packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage
The Quick-Change module is useful when you are running different sample types and need separate sample introduction for each.
Technische Daten
  • Bezeichnung:
    Torch module, quick change, for HF analysis. Includes GemCone high dissolved solids nebuliser, alumina, 2,0 mm Int.Ø injector, quartz 3-slot torch, HF-resistant baffled cyclonic spray chamber, for Optima 4300 V/5300 V/7300 V